Learn More
PD-L1 Antibody (2096C), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB90781MFV450
Product Code. 18543469
  • € 684.00 / 0.10mL
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-L1
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
0.1 mL
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
29126
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry
2096C
Western Blot, Flow Cytometry, Immunohistochemistry
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1, Met1-His239, Accession # Q9EP73