missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (2341A) [Janelia Fluor™ 646], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB90783J
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| PD-L1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 2341A | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1, Phe19-His239, Accession # Q9EP73 | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction