missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (929903) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9078F
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| PD-L1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 929903 | |
| Flow Cytometry, CyTOF-ready | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1, Met1-Thr239, Accession # Q9EP73 | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction