Learn More
PD L1 (CD274) Monoclonal Antibody (OTI14H4), OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812993S
This item is not returnable.
View return policy
Description
PD L1 (CD274) Monoclonal antibody specifically detects PD L1 (CD274) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PD L1 (CD274) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07174, P08138, Q9Z0W1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 18053, 24596, 4804 | |
| -20°C | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI14H4 | |
| Unconjugated | |
| Ngfr | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| Full length human recombinant protein of human Ngfr (NP_002498) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |