missing translation for 'onlineSavingsMsg'
Learn More

PeproTech PD-L1 Fc Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P321BT-50UG

 View more versions of this product

Product Code. 17855653

  • € 284.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: FTVTVPKDLY VVEYGSNMTI ECKFPVEKQL DLAALIVYWE MEDKNIIQFV HGEEDLKVQH SSYRQRARLL KDQLSLGNAA LQITDVKLQD AGVYRCMISY GGADYKRITV KVNAPYNKIN QRILVVDPVT SEHELTCQAE GYPKAEVIWT SSDHQVLSGK TTTTNSKREE KLFNVTSTLR INTTTNEIFY CTFRRLDPEE NHTAELVIPE LPLAHPPNER GGPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PD-L1 Fc. Anti-Human PD-L1 Fc-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human PD-L1 Fc by sandwich ELISA (using 100 μL/well), a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PD-L1 Fc (500-P321) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human PD-L1 Fc. Western Blot: To detect Human PD-L1 Fc by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human PD-L1 Fc is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P321BT-1MG will be provided as 2 x 500 μg

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PD-L1 Fc
Polyclonal
Biotin
Cd274
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Rabbit
Antigen affinity chromatography
RUO
29126
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q9NZQ7
Cd274
CHO cells-derived Recombinant Human PD-L1 Fc.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.