Learn More
Invitrogen™ PD-L1 Monoclonal Antibody (29E.2A3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544052
Description
This antibody recognizes an extracellular epitope of CD274 / PD-L1 (also known as B7-H1), a 40 kDa type I transmembrane protein expressed by dendritic cells, activated T cells, activated monocytes, and in various tissues, above all in heart and skeletal muscle, placenta and lung, and in many cancer cells, including T cell lymphomas, melanomas, and glioblastomas. Functional application: Blocking Immunohistochemistry (Frozen): Acetone fixation.
Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Specifications
| PD-L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NZQ7 | |
| Cd274 | |
| Full length human CD274. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| 29E.2A3 | |
| Unconjugated | |
| Cd274 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 29126 | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.