missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (Z64P2D3*H4), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52408JF549
This item is not returnable.
View return policy
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PD-L2/B7-DC/PDCD1LG2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
| Ovalbumin-conjugated synthetic peptide KAVFFKRHN | |
| 0.1 mL | |
| Apoptosis | |
| 80380 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Z64P2D3*H4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction