missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544938
This item is not returnable.
View return policy
Description
Peptide sequence: MPKGGRKGGH KGRARQYTSP EEIDAQLQAE KQKAREEEEQ KEGGDGAAGD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 86%.
PDGFA-associated protein 1 (PDAP1) is a phosphoprotein that may upregulate the PDGFA-stimulated growth of fibroblasts and also downregulate the mitogenicity of PDGFB. PDGFA in rodents has been shown to bind PDGFA with a low affinity.
Specifications
| PDAP1 | |
| Polyclonal | |
| Unconjugated | |
| PDAP1 | |
| 28 kDa heat- and acid-stable phosphoprotein; HASPP28; heat- and acid-stable phosphoprotein of 28 kDa; kinase substrate HASPP28; PAP; PAP1; Pdap1; PDGFA associated protein 1; PDGFA-associated protein 1; PDGF-associated protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11333 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13442 | |
| PDAP1 | |
| Synthetic peptide directed towards the N-terminal of human PDAP1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction