missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1 Monoclonal Antibody (OTI17G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807207S
This item is not returnable.
View return policy
Description
PDCD1 Monoclonal antibody specifically detects PDCD1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PDCD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62987 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7311 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI17G9 | |
| Unconjugated | |
| UBA52 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| Synthetic peptide corresponding to residues near C-terminus of human UBA52. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction