missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1 Rabbit anti-Human, Clone:RM309, Abnova™
Rabbit monoclonal antibody raised against huamn PDCD1.
Brand: Abnova MAB16936.100uL
This item is not returnable.
View return policy
Description
This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases. [provided by RefSeq]Specifications
| PDCD1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-500) Western Blot (1:100-200) The optimal working dilution should be determined by the end user. |
|
| PDCD1 | |
| Rabbit | |
| 100 μL | |
| This antibody reacts to human PD-1 (Programmed cell death protein 1) (CD279). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM309 | |
| programmed cell death 1 | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| CD279, PD1, SLEB2, hPD-1, hPD-l | |
| A peptide corresponding to N-terminus of human PDCD1. | |
| 5133 | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction