missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD10 Antibody (2A8) [Alexa Fluor« 532], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
PDCD10 Monoclonal antibody specifically detects PDCD10 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | PDCD10 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 2A8 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein |
| Host Species | Mouse |
| Immunogen | programmed cell death 10 - (Uniprot# Q9BUL8) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?