missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD10 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97685APC
This item is not returnable.
View return policy
Description
PDCD10 Polyclonal antibody specifically detects PDCD10 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PDCD10 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
| Produced in rabbits immunized with E. coli-derived Human PDCD10 fragment. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology, Neuroscience, Signal Transduction | |
| 11235 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction