missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD10 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97685UV
This item is not returnable.
View return policy
Description
PDCD10 Polyclonal specifically detects PDCD10 in Human samples. It is validated for Western Blot.
Specifications
| PDCD10 | |
| Polyclonal | |
| Western Blot | |
| apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
| Produced in rabbits immunized with E. coli-derived Human PDCD10 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 11235 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction