missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD2L Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812456
This item is not returnable.
View return policy
Description
Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.Specifications
| PDCD2L | |
| Monoclonal | |
| Unconjugated | |
| PDCD2L | |
| programmed cell death 2 like | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84306 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4H10 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BRP1 | |
| PDCD2L | |
| Full length human recombit protein of human PDCD2L produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction