missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98857-100ul
This item is not returnable.
View return policy
Description
PDCD4 Polyclonal antibody specifically detects PDCD4 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PDCD4 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PDCD4. | |
| 100 μg | |
| Apoptosis | |
| 27250 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction