missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDCD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121697
This item is not returnable.
View return policy
Description
PDCD6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
An increased intracellular Ca2+ concentration induces apoptotic cell death. Transiently elevated Ca2+ concentrations are required for glucocorticoid-mediated and T cell receptor-mediated pathways, leading to T cell apoptosis. ALG-2 (for apoptosis-linked gene 2) is a Ca2+ -binding protein that participates in regulatory events occuring late in the apoptotic program and where several death signals converge. ALG-2 is a protein expressed in normal brain, and to a greater extent in ischemic brain. The ALG-2 protein contains five EF-hand-like motifs and shares homology with members of the penta EF-hand family, which includes Calpain small subunits sorcin and Grancalcin.
Specifications
| PDCD6 | |
| Polyclonal | |
| Unconjugated | |
| Pdcd6 | |
| Alg2; alg-2; ALG-257; Apoptosis-linked gene 2 protein; Apoptosis-linked gene 2 protein homolog; AV299538; MA-3; PDCD6; PEF1B; PMP41; probable calcium-binding protein ALG-2; programmed cell death 6; programmed cell death protein 6; PS2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O75340 | |
| Pdcd6 | |
| Recombinant protein Programmed Cell Death Protein 6. The antigen corresponds to amino acid range 32-191 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction