Learn More
PDCD6IP Mouse anti-Human, Clone: OTI1A4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF507312
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral buding and actin cytoskeleton assembly.Specifications
| PDCD6IP | |
| Monoclonal | |
| Unconjugated | |
| PDCD6IP | |
| AIP1, ALIX, DRIP4, HP95 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10015 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8WUM4 | |
| PDCD6IP | |
| Full length human recombit protein of human PDCD6IP produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.