Learn More
PDCD7 Antibody (46C713.2.1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-56540AF647
Product Code. 18099985
  • € 571.00 / 0.10mL
Description
PDCD7 Monoclonal specifically detects PDCD7 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDCD7
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K
Mouse
Protein G purified
RUO
Primary
Mouse
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
46C713.2.1
Western Blot, Immunocytochemistry/Immunofluorescence
Q8N8D1
PDCD7
The antibody was raised against a synthetic peptide of mouse PDCD-7 protein.
0.1 mL
Apoptosis
10081
Store at 4C in the dark.
IgG1 κ