Learn More
PDCL Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81862JF646
This item is not returnable.
View return policy
Description
PDCL Polyclonal antibody specifically detects PDCL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PDCL | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 646 | |
| DKFZp564M1863, PhLP, phosducin-like, phosducin-like protein | |
| PDCL antibody was raised against a 16 amino acid peptide near the amino terminus of human PDCL. The immunogen is located within the first 50 amino acids of PDCL. Amino Acid Squence: SSEDEDSDHEDKDRGR | |
| 0.1 mL | |
| Signal Transduction | |
| 5082 | |
| Store at 4°C in the dark. | |
| IgG |