missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL3 Antibody (8F9), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42694MFV610
This item is not returnable.
View return policy
Description
PDCL3 Monoclonal antibody specifically detects PDCL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| PDCL3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 8F9 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| HTPHLP, IAP-associated factor VIAF1, MGC3062, PHLP3, phosducin-like 3, phosducin-like protein 3, PhPL3, VIAF, VIAF-1, VIAF1PHLP2A, Viral IAP-associated factor 1 | |
| Recombinant human PDCL3 (1-239aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis | |
| 79031 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction