missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL3 Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809771
This item is not returnable.
View return policy
Description
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.Specifications
| PDCL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8BVF2, Q9H2J4 | |
| PDCL3 | |
| Full length human recombit protein of human PDCL3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A6 | |
| Unconjugated | |
| PDCL3 | |
| HTPHLP, PHLP2A, PHLP3, VIAF, VIAF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 68833, 79031 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction