missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL3 Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809771S
This item is not returnable.
View return policy
Description
PDCL3 Monoclonal antibody specifically detects PDCL3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PDCL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q53EU6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84803 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A6 | |
| Unconjugated | |
| GPAT3 | |
| 1110061A19Rik; C80025; HTPHLP; IAP-associated factor VIAF1; PDCL3; PhLP2A; PHLP3; phosducin like 3; phosducin-like 3; phosducin-like protein 3; PhPL3; VIAF; VIAF1; VIAF-1; Viral IAP-associated factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 41-131 of human AGPAT9 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction