missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PDE10A Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Specifications
Specifications
| Gene ID (Entrez) | 10846 |
| Species | Human |
| Purity | 99.9% |
| Concentration | 2.5 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB300-645 |
| Gene Alias | cAMP and cAMP-inhibited cGMP 3 '-5 '-cyclic phosphodiesterase 10 A, dJ416F21.1 (phosphodiesterase 10 A), EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ11894, FLJ25677, HSPDE10A, phosphodiesterase 10 A, phosphodiesterase 10A1 (PDE10A1) |
| Gene Symbol | PDE10A |
| Quantity | 0.25 mg |
| Storage Buffer | Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction