missing translation for 'onlineSavingsMsg'
Learn More

PDE10A Mouse anti-Human, Clone: OTI1C9, liquid, TrueMAB™

Product Code. 17591961
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17591961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17591961 Supplier Origene Technologies Supplier No. TA500625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.

Specifications

Antigen PDE10A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C9
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDE10A
Gene Accession No. Q9Y233
Gene Alias ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19
Gene Symbols PDE10A
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PDE10A expression vector
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10846
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.