missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560267
This item is not returnable.
View return policy
Description
Immunogen sequence: CGDFNSTPST GMYHFVINGS IPEDHEDWAS NGEEERCNMS LTHFFKLKSA CGEPAYTNYV GGFHGCLDYI FIDLNALEVE QVIPLPSHEE VTT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 97%.
PDE12 is an enzyme that cleaves 2',5'-phosphodiester bond linking adenosines of the 5'-triphosphorylated oligoadenylates, triphosphorylated oligoadenylates referred as 2-5A modulates the 2-5A system. This enzyme degraded triphosphorylated 2-5A to produce AMP and ATP. It also cleaves 3',5'-phosphodiester bond of oligoadenylates and plays a role as a negative regulator of the 2-5A system that is one of the major pathways for antiviral and antitumor functions induced by interferons (IFNs). Suppression of this enzyme induces reduction of viral replication in Hela cells, thus counteracting the antiviral pathway probably by inhibiting the 2-5A system.
Specifications
| PDE12 | |
| Polyclonal | |
| Unconjugated | |
| PDE12 | |
| 2',5'-phosphodiesterase 12; 2',5'-phosphodiesterase 12-like protein; 2'-PDE; 2-PDE; 2'-phosphodiesterase; E430028B21Rik; fc58c05; fi42f03; LOC558284 protein; LOW QUALITY PROTEIN: 2',5'-phosphodiesterase 12; LRRGT00074; Mitochondrial deadenylase; PDE12; phosphodiesterase 12; RGD1310975; wu:fc58c05; wu:fi42f03; zgc:136374 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6L8Q7 | |
| PDE12 | |
| Recombinant protein corresponding to Human PDE12. Recombinant protein control fragment (Product #RP-100240). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction