missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1A Antibody (OTI7D5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73277UV
This item is not returnable.
View return policy
Description
PDE1A Monoclonal specifically detects PDE1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PDE1A | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDE1A | |
| Recombinant protein fragment corresponding to amino acids 259-545 of human PDE1A (NP_005010) produced in E.coli. | |
| 0.1 mL | |
| 5136.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI7D5 | |
| Western Blot, Immunohistochemistry | |
| 3′-5′ cyclic nucleotide phosphodiesterase, 61 kDa Cam-PDE, calcium/calmodulin-dependent 3′-5′-cyclic nucleotide phosphodiesterase 1A, calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, calmodulin-dependent phosphodiesterase, Cam-PDE 1A, EC 3.1.4, EC 3.1.4.17, hCam-1, HCAM1, HSPDE1A, MGC26303, phosphodiesterase 1A, calmodulin-dependent, phosphodiesterase-1A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction