missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 13289478
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13289478 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13289478 Supplier Invitrogen™ Supplier No. PA522184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (93%), Rat (93%), Dog (89%), Rhesus Monkey (97%), Chimpanzee (100%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cyclic nucleotide phosphodiesterases (PDEs) play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Ca(2+)/calmodulin (see CALM1)that are activated by calmodulin in the presence of Ca(2+) (Michibata et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PDE1A
Gene Accession No. P54750, Q61481
Gene Alias 3'-RACE clone 8 phosphodiesterase 1A; 3-RACE clone 8 phosphodiesterase 1A; 3'-RACEclone8phosphodiesterase1A; 3-RACEclone8phosphodiesterase1A; 61 kDa Cam-PDE; AI987702; AW125737; calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; cam-PDE 1A; CAM-PDE-1A; HCAM1; hCam-1; HS MGC26303; HSPDE1A; MGC26303; Pde1a; phosphodiesterase 1A; phosphodiesterase 1A, calmodulin-dependent; phosphodiesterase-1A
Gene Symbols PDE1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDE1A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18573, 5136, 81529
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.