missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1B Antibody (OTI6E4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73278AF594
This item is not returnable.
View return policy
Description
PDE1B Monoclonal specifically detects PDE1B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| PDE1B | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PDE1B | |
| Full length recombinant protein of human PDE1B (NP_000915) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6E4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 63 kDa Cam-PDE, calcium/calmodulin-dependent 3′-5′-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Signal Transduction | |
| 5153.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction