missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00157-50ul
This item is not returnable.
View return policy
Description
PDE1C Polyclonal antibody specifically detects PDE1C in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PDE1C | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| calmodulin-dependent (70kD), EC 3.1.4, phosphodiesterase 1C, calmodulin-dependent 70kDa | |
| Produced in rabbits immunized with purified, recombinant Human PDE1C (Uniprot#: Q8TAE4; Met1-Glu634) | |
| 50 μg | |
| Signal Transduction | |
| 5137 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction