missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE2A Antibody (SR1040), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22378-50ul
This item is not returnable.
View return policy
Description
PDE2A Monoclonal antibody specifically detects PDE2A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PDE2A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1040 | |
| Western Blot 1:500-1:2000 | |
| cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
| A synthesized peptide derived from human PDE2A (Uniprot #: O00408) | |
| 50 μg | |
| Signal Transduction | |
| 5138 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction