missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT8L2 Monoclonal Antibody (OTI1D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505304
This item is not returnable.
View return policy
Description
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.Specifications
| CCT8L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96SF2 | |
| CCT8L2 | |
| Full length human recombit protein of human CCT8L2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1D12 | |
| Unconjugated | |
| CCT8L2 | |
| CESK1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 150160 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction