missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA131130
This item is not returnable.
View return policy
Description
PA1-31130 detects PDE4A8 from rat, mouse samples.
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.
Specifications
| PDE4A | |
| Polyclonal | |
| Unconjugated | |
| Pde4a | |
| cAMP-specific 3',5'-cyclic phosphodiesterase 4A; cAMP-specific phosphodiesterase PDE4A; cyclic AMP specific phosphodiesterase PDE4A5A; cyclic AMP-specific phosphodiesterase OCPDE4A1A; D9Ertd60e; DPDE2; dunce; LOC100009247; PDE4; PDE46; Pde4a; PHOSA; phosphodiesterase 4A; phosphodiesterase 4A, cAMP specific; phosphodiesterase 4A, cAMP-specific; phosphodiesterase 4A, cAMP-specific (dunce; phosphodiesterase 4A, cAMP-specific (phosphodiesterase E2 dunce homolog, Drosophila); phosphodiesterase E2 dunce homolog, Drosophila; phosphodiesterase isozyme 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18577, 25638, 5141 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| Tris glycine with 0.5% BSA, 30% glycerol and 0.02% Sodium azide; pH 7.4-7.9 | |
| O89084, P27815, P54748 | |
| Pde4a | |
| Synthetic peptide specific only for PDE4A8 protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction