Learn More
PDE4B Monoclonal Antibody (OTI2B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA503446
Encompass_Preferred
 View more versions of this product
Product Code. 17539301
  • € 540.87 / 100µL
Description
This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
PDE4B
Monoclonal
0.53 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q07343
PDE4B
Full length human recombit protein of human PDE4B produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI2B10
Unconjugated
PDE4B
DPDE4, PDEIVB
Mouse
Affinity Chromatography
RUO
5142
-20° C, Avoid Freeze/Thaw Cycles