Learn More
PDE4B Mouse anti-Human, Mouse, Clone: OTI1D12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503471
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| PDE4B | |
| Monoclonal | |
| Unconjugated | |
| PDE4B | |
| DPDE4, PDEIVB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18578, 5142 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q07343 | |
| Pde4b | |
| Full length human recombit protein of human PDE4B produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.