missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE4C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549996
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PDE4C protein.
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases.
Specifications
| PDE4C | |
| Polyclonal | |
| Unconjugated | |
| PDE4C | |
| cAMP-specific 3',5'-cyclic phosphodiesterase 4C; cAMP-specific 35-cyclic phosphodiesterase 4C; CN4C; DPDE1; dunce; E130301F19Rik; MGC126222; PDE21; Pde4c; phosphodiesterase 4C; phosphodiesterase 4C, cAMP specific; phosphodiesterase 4C, cAMP-specific; phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase E1 dunce homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110385, 5143 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q08493, Q3UEI1 | |
| PDE4C | |
| Synthesized peptide derived from internal of human PDE4C. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction