missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6D Mouse anti-Human, Unconjugated, Clone: 1H4B6, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | PDE6D |
|---|---|
| Clone | 1H4B6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860266
|
Proteintech
67565-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850266
|
Proteintech
67565-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PDE6D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDE6D. This antibody recognizes human, mouse, rat antigen. The PDE6D antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PDE6D | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 5147 | |
| Primary |
| 1H4B6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title