missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6G Antibody (OTI2F5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73298PE
This item is not returnable.
View return policy
Description
PDE6G Monoclonal antibody specifically detects PDE6G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PDE6G | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2F5 | |
| Western Blot, Immunohistochemistry | |
| DKFZp686C0587, GMP-PDE gamma, MGC125749, PDEG, phosphodiesterase 6G, cGMP-specific, rod, gamma, retinal rod rhodopsin-sensitive cGMP 3'-5'-cyclic phosphodiesterase subunit gamma, rod cG-PDE G, RP57 | |
| Full length human recombinant protein of human PDE6G(NP_002593) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 5148 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction