missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6G Monoclonal Antibody (OTI2F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503736S
This item is not returnable.
View return policy
Description
PDE6G Monoclonal antibody specifically detects PDE6G in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PDE6G | |
| Monoclonal | |
| 0.72 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TB37 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80224 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F5 | |
| Unconjugated | |
| Nubpl | |
| GMP-PDE gamma; p3AT; PDE6G; PDEG; phosphodiesterase 6G; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase, cyclic GMP (rod receptor), gamma polypeptide; retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma; rod cG-PDE G; RP57; unnamed protein product | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction