missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE8B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583361

Product Code. 16300325

  • € 534.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FVSLKKLCCT TDNNKQIHKI HRDSGDNSQT EPHSFRYKNR RKESIDVKSI SSRGSDAPSL QNRRYPS Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze hydrolysis of the 5-prime,3-prime-cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. Mammalian PDEs have been grouped into several families based on substrate affinities, inhibitor sensitivities, mode of regulation, and amino acid sequence homologies. The PDE8 family contains high-affinity cAMP-specific, IBMX (3-isobutyl-1-methyl-xanthine)-insensitive PDEs, such as PDE8B. All PDEs share a conserved C-terminal catalytic region and a variable N-terminal domain that presumably accounts for the distinctive regulatory properties unique to the individual families.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PDE8B
Polyclonal
Unconjugated
PDE8B
3',5' cyclic nucleotide phosphodiesterase 8B; ADSD; B230331L10Rik; C030047E14Rik; cAMP-specific cyclic nucleotide phosphodiesterase PDE8; cell proliferation-inducing gene 22 protein; FLJ11212; High affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8B; hsPDE8B; Pde8b; phosphodiesterase 8B; PIG22; PPNAD3; RNPDE8B
Rabbit
Antigen affinity chromatography
RUO
8622
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95263
PDE8B
Recombinant protein corresponding to Human PDE8B. Recombinant protein control fragment (Product #RP-96876).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.