missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE9A Polyclonal Antibody

Product Code. 11511633
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11511633 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11511633 Supplier Invitrogen™ Supplier No. PA131148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-31148 detects PDE9A from rat, human, and mouse samples.

The protein encoded by this gene catalyzes the hydrolysis of cAMP and cGMP to their corresponding monophosphates. The encoded protein plays a role in signal transduction by regulating the intracellular concentration of these cyclic nucleotides. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE9A
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation Tris glycine with 0.5% BSA, 30% glycerol and 0.02% Sodium azide; pH 7.4-7.9
Gene PDE9A
Gene Accession No. O70628, O76083, Q8QZV1
Gene Alias CGMP-specific 3',5'-cyclic phosphodiesterase type 9; FLJ90181; high affinity cGMP-specific 3',5'-cyclic phosphodiesterase 9A; HSPDE9A2; Pde8b; PDE9A; PDE9A1; phosphodiesterase 9A; phosphodiesterase PDE9A21
Gene Symbols PDE9A
Host Species Rabbit
Immunogen Synthetic cyclic peptide common to all PDE9A variants.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18585, 191569, 5152
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.