missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE9A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572944
This item is not returnable.
View return policy
Description
PDE9A Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, IP
The protein encoded by this gene catalyzes the hydrolysis of cAMP and cGMP to their corresponding monophosphates. The encoded protein plays a role in signal transduction by regulating the intracellular concentration of these cyclic nucleotides. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
| PDE9A | |
| Polyclonal | |
| Unconjugated | |
| PDE9A | |
| CGMP-specific 3',5'-cyclic phosphodiesterase type 9; FLJ90181; high affinity cGMP-specific 3',5'-cyclic phosphodiesterase 9A; HSPDE9A2; Pde8b; PDE9A; PDE9A1; phosphodiesterase 9A; phosphodiesterase PDE9A21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18585, 191569, 5152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 30% glycerol and 0.05% sodium azide | |
| O70628, O76083, Q8QZV1 | |
| PDE9A | |
| Sequence from the C-terminal region of PDE9A. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction