missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98168AF405
This item is not returnable.
View return policy
Description
PDF Polyclonal antibody specifically detects PDF in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
| Produced in rabbits immunized with E. coli-derived Human PDF fragment. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction | |
| 64146 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction