missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Antibody (OTI3F3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73301
This item is not returnable.
View return policy
Description
PDF Monoclonal specifically detects PDF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
| EC 3.5.1.88, GDF15, MIC1, PDF1A, peptide deformylase (mitochondrial), peptide deformylase, mitochondrial, peptide deformylase-like protein, PLAB, Polypeptide deformylase | |
| Mouse | |
| 19.4 kDa | |
| 100 μg | |
| 64146 | |
| Human | |
| Lyophilized |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| OTI3F3 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| Full length human recombinant protein of human PDF (NP_071736) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction