missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-B Antibody (MM0014-5F66), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60980B
This item is not returnable.
View return policy
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PDGF-B | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PDGFB | |
| Human recombinant PDGF-B | |
| 0.1 mL | |
| Primary | |
| This antibody shows approximately some degree (40%) cross-reactivity with recombinant PDGF-AB and no cross-reactivity with recombinant PDGF-AA. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0014-5F66 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
| 5155 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction