missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-C Antibody (004), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89373APC
This item is not returnable.
View return policy
Description
PDGF-C Monoclonal antibody specifically detects PDGF-C in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| PDGF-C | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry | |
| 004 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGF-C (Accession#: NP_057289.1; Val235-Gly345). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 56034 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction