missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-C Antibody (5F12), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21956C
This item is not returnable.
View return policy
Description
PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for Western Blot.
Specifications
| PDGF-C | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDGFC | |
| Human recombinant PDGF-C | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 5F12 | |
| Western Blot | |
| FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication | |
| 56034 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction