Learn More
Novus Biologicals™ PDGF R beta Antibody (IMC-2C5), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28360-50ug
 View more versions of this product
Product Code. 30047862
  • € 213.00 / 50µg
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGF R beta
Monoclonal
Unconjugated
P09619
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
IMC-2C5
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
PDGFRB/ CD140b
50 μg
Oncogenes, Phospho Specific, Protein Kinase
5159