missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PDGF R beta Antibody Pair [HRP]

Product Code. p-7183257 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18225286

Brand: Novus Biologicals™ NBP27948115PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human PDGF R beta.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human PDGF R beta.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Gene ID (Entrez) 5159
Gene Symbol PDGFRB
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL of mouse anti-PDGF R beta monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.3 mg/mL of mouse anti-PDGF R beta monoclonal antibody.
  • Standard: Each vial contains 40 ng of recombinant PDGF R beta.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Oncogenes, Phospho Specific, Protein Kinase
    Storage Requirements Storage is content dependent.
    Target PDGF R beta
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to a working concentration of 0.8 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.