missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFA Antibody (MM0013-6D35), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60979CL1
This item is not returnable.
View return policy
Description
PDGFA Monoclonal antibody specifically detects PDGFA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PDGFA | |
| Monoclonal | |
| CoraFluor 1 | |
| PDGF-1, PDGF1PDGF A-chain, PDGF-A, Platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor alpha polypeptidePDGF subunit A, platelet-derived growth factor subunit A | |
| Human recombinant PDGFA | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5154 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| MM0013-6D35 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction