missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFD Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503700S
This item is not returnable.
View return policy
Description
PDGFD Monoclonal antibody specifically detects PDGFD in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PDGFD | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| 1110003I09Rik; Iegf; Iris-expressed growth factor; MSTP036; Pdgfd; PDGF-D; PDGFD latent form; PDGFD receptor-binding form; platelet derived growth factor D; platelet-derived growth factor D; Platelet-derived growth factor D, latent form; Platelet-derived growth factor D, receptor-binding form; platelet-derived growth factor, D polypeptide; rSCDGF-B; SCDGFB; SCDGF-B; spinal cord derived growth factor B; spinal cord-derived growth factor B; spinal cord-derived growth factor-B; spinal-cord derived growth factor-B protein; UNQ1899/PRO4345 | |
| Full length human recombinant protein of human KIAA0495 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| TP73-AS1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57212 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction